COMH4211AED0 Combi-Blocks, Inc.

3-Bromo-4-(2,2,2-trifluoroethoxy)nitrobenzene

  • MDL number MFCD28154866
  • Cas Number : 710351-85-2
  • Molecular Weight : 300.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2557

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2557
Availability details for
Quantity
  • Availability
  • Add to Cart