COMH4211AED2 Combi-Blocks, Inc.

1-Bromo-4-(3-methoxypropyl)benzene

  • MDL number MFCD27945854
  • Cas Number : 166960-00-5
  • Molecular Weight : 229.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2560

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2560
Availability details for
Quantity
  • Availability
  • Add to Cart