COMH4211AED3 Combi-Blocks, Inc.

1-(4-Pyrrolidin-1-ylmethyl-phenyl)-ethanone

  • MDL number MFCD10000740
  • Cas Number : 149704-65-4
  • Molecular Weight : 203.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2561

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2561
Availability details for
Quantity
  • Availability
  • Add to Cart