COMH4211AED4 Combi-Blocks, Inc.

4-(2-Fluoro-4-nitro-phenoxy)-benzaldehyde

  • MDL number MFCD13240919
  • Cas Number : 875700-55-3
  • Molecular Weight : 261.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2562

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2562
Availability details for
Quantity
  • Availability
  • Add to Cart