COMH4211AED6 Combi-Blocks, Inc.

5-Diethylcarbamoyl-pentanoic acid

  • MDL number MFCD12172658
  • Cas Number : 91017-35-5
  • Molecular Weight : 201.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2566

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2566
Availability details for
Quantity
  • Availability
  • Add to Cart