COMH4211AEDC Combi-Blocks, Inc.

N-(1,3-Benzodioxol-5-ylmethyl)urea

  • MDL number MFCD05854325
  • Cas Number : 65609-28-1
  • Molecular Weight : 194.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2584

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2584
Availability details for
Quantity
  • Availability
  • Add to Cart