COMH4211AEDD Combi-Blocks, Inc.

(3-Bromo-4-ethoxy-5-methoxyphenyl)methanol

  • MDL number MFCD08277372
  • Cas Number : 898693-07-7
  • Molecular Weight : 261.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2585

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2585
Availability details for
Quantity
  • Availability
  • Add to Cart