COMH4211AEDE Combi-Blocks, Inc.

4-Fluoro-2-methyl-5-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-benzoic acid benzyl ester

  • MDL number MFCD31579456
  • Molecular Weight : 370.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2586

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2586
Availability details for
Quantity
  • Availability
  • Add to Cart