COMH4211AEDF Combi-Blocks, Inc.

tert-Butyl 2-(2-iodophenoxy)acetate

  • MDL number MFCD26403927
  • Cas Number : 1229805-95-1
  • Molecular Weight : 334.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2588

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2588
Availability details for
Quantity
  • Availability
  • Add to Cart