COMH4211AEE0 Combi-Blocks, Inc.

(4-Isopropyl-phenyl)-phenyl-methanone

  • MDL number MFCD00096246
  • Cas Number : 18864-76-1
  • Molecular Weight : 224.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2589

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2589
Availability details for
Quantity
  • Availability
  • Add to Cart