COMH4211AEE1 Combi-Blocks, Inc.

2-Chloro-4-phenylpyrimidin-5-amine

  • MDL number MFCD31557911
  • Cas Number : 1812201-89-0
  • Molecular Weight : 205.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2590

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2590
Availability details for
Quantity
  • Availability
  • Add to Cart