COMH4211AEE2 Combi-Blocks, Inc.

5-Bromo-2-(2,2,2-trifluoro-ethoxy)-benzoic acid methyl ester

  • MDL number MFCD31381723
  • Cas Number : 2205470-20-6
  • Molecular Weight : 313.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2593

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2593
Availability details for
Quantity
  • Availability
  • Add to Cart