COMH4211AEE4 Combi-Blocks, Inc.

3-Bromo-2,6-dimethylbenzaldehyde

  • MDL number MFCD28637585
  • Cas Number : 106965-52-0
  • Molecular Weight : 213.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2595

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2595
Availability details for
Quantity
  • Availability
  • Add to Cart