COMH4211AEE7 Combi-Blocks, Inc.

4-(2,2-Dimethyl-1,3-dioxolan-4-ylmethoxy)-2-trifluoromethyl-phenylamine

  • MDL number MFCD31614003
  • Cas Number : 873455-96-0
  • Molecular Weight : 291.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2602

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2602
Availability details for
Quantity
  • Availability
  • Add to Cart