COMH4211AEE8 Combi-Blocks, Inc.

6-Methyl-[1,2,4]triazolo[4,3-b]pyridazine-3-thiol

  • MDL number MFCD23725584
  • Cas Number : 89418-40-6
  • Molecular Weight : 166.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2603

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2603
Availability details for
Quantity
  • Availability
  • Add to Cart