COMH4211AEE9 Combi-Blocks, Inc.

Ethyl 4-(3-bromophenoxy)butanoate

  • MDL number MFCD20483435
  • Cas Number : 157245-84-6
  • Molecular Weight : 287.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2604

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2604
Availability details for
Quantity
  • Availability
  • Add to Cart