COMH4211AEED Combi-Blocks, Inc.

4-[2-(4-Oxopiperidin-1-yl)ethyl]piperidine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD31557861
  • Cas Number : 918884-45-4
  • Molecular Weight : 310.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 4-(2-(4-oxopiperidin-1-yl)ethyl)piperidine-1-carboxylate
Inchi Code 1S/C17H30N2O3/c1-17(2,3)22-16(21)19-12-5-14(6-13-19)4-9-18-10-7-15(20)8-11-18/h14H,4-13H2,1-3H3
Inchi Key PSYIDVUTCBODQT-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2611

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2611
Availability details for
Quantity
  • Availability
  • Add to Cart