COMH4211AEEE Combi-Blocks, Inc.

1-(2-Isopropoxy-5-methyl-phenyl)-ethylamine

  • MDL number MFCD11053242
  • Cas Number : 1048916-98-8
  • Molecular Weight : 193.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2612

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2612
Availability details for
Quantity
  • Availability
  • Add to Cart