COMH4211AEF0 Combi-Blocks, Inc.

4-(4-Bromo-phenyl)-piperazine-1-carboxylic acid methylamide

  • MDL number MFCD31583316
  • Cas Number : 2270907-72-5
  • Molecular Weight : 298.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2617

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2617
Availability details for
Quantity
  • Availability
  • Add to Cart