COMH4211AEF3 Combi-Blocks, Inc.

1-(4-Amino-phenyl)-3-benzyl-urea

  • MDL number MFCD12414527
  • Cas Number : 184177-11-5
  • Molecular Weight : 241.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2623

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2623
Availability details for
Quantity
  • Availability
  • Add to Cart