COMH4211AEF4 Combi-Blocks, Inc.

4-Isopropoxy-3-nitrobenzoic acid

  • MDL number MFCD09810083
  • Cas Number : 156629-52-6
  • Molecular Weight : 225.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-isopropoxy-3-nitrobenzoic acid
Inchi Code 1S/C10H11NO5/c1-6(2)16-9-4-3-7(10(12)13)5-8(9)11(14)15/h3-6H,1-2H3,(H,12,13)
Inchi Key YRJMQBFZZKIYDN-UHFFFAOYSA-N

Description

Purity 96%
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2625

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2625
Availability details for
Quantity
  • Availability
  • Add to Cart