COMH4211AEF5 Combi-Blocks, Inc.

4-Cyclohexylmethoxy-phenol

  • MDL number MFCD11181760
  • Cas Number : 215865-15-9
  • Molecular Weight : 206.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2630

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2630
Availability details for
Quantity
  • Availability
  • Add to Cart