COMH4211AEF8 Combi-Blocks, Inc.

4-[2-Methoxy-5-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-benzenesulfonylamino]-piperidine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD31583303
  • Cas Number : 1628017-41-3
  • Molecular Weight : 496.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2640

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2640
Availability details for
Quantity
  • Availability
  • Add to Cart