COMH4211AEF9 Combi-Blocks, Inc.

4-(2-Methoxy-ethoxy)-3,5-dimethyl-benzylamine

  • MDL number MFCD11192011
  • Cas Number : 1039317-01-5
  • Molecular Weight : 209.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2641

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2641
Availability details for
Quantity
  • Availability
  • Add to Cart