COMH4211AEFA Combi-Blocks, Inc.

5-Amino-4-fluoro-2-methyl-n-propyl-benzamide

  • MDL number MFCD31579292
  • Cas Number : 1881414-75-0
  • Molecular Weight : 210.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2643

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2643
Availability details for
Quantity
  • Availability
  • Add to Cart