COMH4211AEFC Combi-Blocks, Inc.

4-(4-Ethoxy-2-fluoro-phenyl)-piperidine hydrochloride

  • MDL number MFCD31583396
  • Molecular Weight : 259.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2652

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2652
Availability details for
Quantity
  • Availability
  • Add to Cart