COMH4211AEFE Combi-Blocks, Inc.

N-[(Phenylmethoxy)carbonyl]-l-norvalyl-l-leucinamide

  • MDL number MFCD00026505
  • Cas Number : 196600-89-2
  • Molecular Weight : 363.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2656

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2656
Availability details for
Quantity
  • Availability
  • Add to Cart