COMH4211AEFF Combi-Blocks, Inc.

5-Bromo-2-(4-methyl-piperazin-1-yl)-nicotinic acid tert-butyl ester

  • MDL number MFCD31557807
  • Cas Number : 2206265-67-8
  • Molecular Weight : 356.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2659

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2659
Availability details for
Quantity
  • Availability
  • Add to Cart