COMH4211AF02 Combi-Blocks, Inc.

2-(Cyclopropylmethoxy)-5-hydroxybenzoic acid

  • MDL number MFCD17000211
  • Cas Number : 1243367-94-3
  • Molecular Weight : 208.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2665

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2665
Availability details for
Quantity
  • Availability
  • Add to Cart