COMH4211AF03 Combi-Blocks, Inc.

1-Bromo-4-[(5-bromopentyl)oxy]benzene

  • MDL number MFCD00596649
  • Cas Number : 91130-83-5
  • Molecular Weight : 322.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2668

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2668
Availability details for
Quantity
  • Availability
  • Add to Cart