COMH4211AF05 Combi-Blocks, Inc.

[3,5-Dimethyl-4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-pyrazol-1-yl]-acetic acid benzyl ester

  • MDL number MFCD31583227
  • Molecular Weight : 370.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2677

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2677
Availability details for
Quantity
  • Availability
  • Add to Cart