COMH4211AF06 Combi-Blocks, Inc.

4-(Methoxyamino-methyl)-benzoic acid methyl ester

  • MDL number MFCD21100473
  • Cas Number : 543731-23-3
  • Molecular Weight : 195.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2678

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2678
Availability details for
Quantity
  • Availability
  • Add to Cart