COMH4211AF07 Combi-Blocks, Inc.

1-(3-Bromo-benzyl)-1h-pyrazol-4-ol

  • MDL number MFCD31561492
  • Cas Number : 1602366-61-9
  • Molecular Weight : 253.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2679

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2679
Availability details for
Quantity
  • Availability
  • Add to Cart