COMH4211AF08 Combi-Blocks, Inc.

Butanedioic acid, 1-(1,1-dimethylethyl) 4-(2,5-dioxo-1-pyrrolidinyl) ester

  • MDL number MFCD30290246
  • Cas Number : 138833-93-9
  • Molecular Weight : 271.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2680

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2680
Availability details for
Quantity
  • Availability
  • Add to Cart