COMH4211AF0A Combi-Blocks, Inc.

[1-(2-Bromo-4-methyl-phenyl)-ethyl]-carbamic acid tert-butyl ester

  • MDL number MFCD31539539
  • Cas Number : 2167451-57-0
  • Molecular Weight : 314.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2686

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2686
Availability details for
Quantity
  • Availability
  • Add to Cart