COMH4211AF0B Combi-Blocks, Inc.

tert-Butyl 1-[(6-bromopyridin-2-yl)methyl]piperidin-4-ylcarbamate

  • MDL number MFCD09028518
  • Cas Number : 303763-37-3
  • Molecular Weight : 370.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2688

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2688
Availability details for
Quantity
  • Availability
  • Add to Cart