COMH4211AF0F Combi-Blocks, Inc.

4-[2-(4-Bromo-phenyl)-2-methoxycarbonyl-ethyl]-benzoic acid tert-butyl ester

  • MDL number MFCD29071497
  • Cas Number : 1207989-81-8
  • Molecular Weight : 419.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2693

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2693
Availability details for
Quantity
  • Availability
  • Add to Cart