COMH4211AF12 Combi-Blocks, Inc.

2-(Cyclopentanesulfonyl)benzoic acid

  • MDL number MFCD09928367
  • Cas Number : 1016722-72-7
  • Molecular Weight : 254.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2697

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2697
Availability details for
Quantity
  • Availability
  • Add to Cart