COMH4211AF13 Combi-Blocks, Inc.

N-(4-Chlorophenyl)-4-oxopiperidine-1-carboxamide

  • MDL number MFCD11128334
  • Cas Number : 1019388-65-8
  • Molecular Weight : 252.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2700

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2700
Availability details for
Quantity
  • Availability
  • Add to Cart