COMH4211AF15 Combi-Blocks, Inc.

(3S)-N-(Cyclopropylmethyl)piperidine-3-carboxamide

  • MDL number MFCD24052250
  • Cas Number : 1567924-96-2
  • Molecular Weight : 182.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (S)-N-(cyclopropylmethyl)piperidine-3-carboxamide
Inchi Code 1S/C10H18N2O/c13-10(12-6-8-3-4-8)9-2-1-5-11-7-9/h8-9,11H,1-7H2,(H,12,13)/t9-/m0/s1
Inchi Key LISFHWSTVXRTSU-VIFPVBQESA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2707

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2707
Availability details for
Quantity
  • Availability
  • Add to Cart