COMH4211AF18 Combi-Blocks, Inc.

tert-Butyl (2,5-dioxopyrrolidin-1-yl) adipate

  • MDL number MFCD31381621
  • Cas Number : 402958-62-7
  • Molecular Weight : 299.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2714

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2714
Availability details for
Quantity
  • Availability
  • Add to Cart