COMH4211AF19 Combi-Blocks, Inc.

4-(Methoxymethyl)-3-nitrobenzoic acid

  • MDL number MFCD12173015
  • Cas Number : 869186-74-3
  • Molecular Weight : 211.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2720

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2720
Availability details for
Quantity
  • Availability
  • Add to Cart