COMH4211AF1A Combi-Blocks, Inc.

1-(3-Chloro-4-fluorobenzyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1h-pyrazole

  • MDL number MFCD28718210
  • Cas Number : 1604036-60-3
  • Molecular Weight : 336.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2722

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2722
Availability details for
Quantity
  • Availability
  • Add to Cart