COMH4211AF1D Combi-Blocks, Inc.

2-Chloro-5-(2-fluorophenyl)pyrimidine

  • MDL number MFCD14698946
  • Cas Number : 1048338-45-9
  • Molecular Weight : 208.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-chloro-5-(2-fluorophenyl)pyrimidine
Inchi Code 1S/C10H6ClFN2/c11-10-13-5-7(6-14-10)8-3-1-2-4-9(8)12/h1-6H
Inchi Key ZTHDGOABMVTBFB-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2733

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2733
Availability details for
Quantity
  • Availability
  • Add to Cart