COMH4211AF1F Combi-Blocks, Inc.

1-(2-Methoxyethyl)-1h-pyrazol-4-ol

  • MDL number MFCD26129298
  • Cas Number : 1601962-79-1
  • Molecular Weight : 142.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2739

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2739
Availability details for
Quantity
  • Availability
  • Add to Cart