COMH4211AF20 Combi-Blocks, Inc.

1-(3-Bromo-4-fluoro-phenyl)-4-cyclopropyl-butane-1,3-dione

  • MDL number MFCD30536351
  • Cas Number : 2065223-86-9
  • Molecular Weight : 299.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2743

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2743
Availability details for
Quantity
  • Availability
  • Add to Cart