COMH4211AF22 Combi-Blocks, Inc.

tert-Butyl 4-(3-hydroxyphenoxy)piperidine-1-carboxylate

  • MDL number MFCD23110530
  • Cas Number : 528861-46-3
  • Molecular Weight : 293.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2755

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2755
Availability details for
Quantity
  • Availability
  • Add to Cart