COMH4211AF23 Combi-Blocks, Inc.

2-(2-Bromophenyl)-n-(cyclopropylmethyl)acetamide

  • MDL number MFCD31539542
  • Cas Number : 1311779-40-4
  • Molecular Weight : 268.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2757

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2757
Availability details for
Quantity
  • Availability
  • Add to Cart