COMH4211AF24 Combi-Blocks, Inc.

7-Iodo-1-heptanol

  • MDL number MFCD30469163
  • Cas Number : 89940-48-7
  • Molecular Weight : 242.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2760

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2760
Availability details for
Quantity
  • Availability
  • Add to Cart