COMH4211AF26 Combi-Blocks, Inc.

3,5-Dimethyl-1-(2-methyl-benzyl)-4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-1h-pyrazole

  • MDL number MFCD31583212
  • Cas Number : 2246835-74-3
  • Molecular Weight : 326.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2762

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2762
Availability details for
Quantity
  • Availability
  • Add to Cart